Learn More
Grainger Legend Plate: 22 mm Panel Mounting Size, Round, 2 3/8 in Overall Dia, For Series A22, "Emergency Stop"
Catalog No. NC3888203
Encompass_Preferred
Plates for push button legends feature printed text to provide information about a push button's function. These plates insert between a push button's head and its body, or the head and the lighting module of the button assembly.
Catalog No. NC3888203 Supplier Grainger Supplier No. 2LCC3
Plates for push button legends feature printed text to provide information about a push button's function. These plates insert between a push button's head and its body, or the head and the lighting module of the button assembly.