Learn More
Grainger LegendPlate HandAuto Wht/Blk Rectangular
Catalog No. 502753582
Encompass_Preferred
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter pushbutton switch with a legend plate. It has a plastic housing and is rated for 10 amps at 250 volts AC. The pushbutton has a silver colored metal actuator and includes a legend plate that can be customized with text or symbols. It mounts into a panel cutout and has quick connect terminals for easy wiring. This pushbutton switch is commonly used in industrial control panels and machinery.
Catalog No. 50-275-3582 Supplier Grainger Supplier No. 14A107
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter pushbutton switch with a legend plate. It has a plastic housing and is rated for 10 amps at 250 volts AC. The pushbutton has a silver colored metal actuator and includes a legend plate that can be customized with text or symbols. It mounts into a panel cutout and has quick connect terminals for easy wiring. This pushbutton switch is commonly used in industrial control panels and machinery.