Learn More
Grainger LegendPlate High White/Black Rectangular
Catalog No. 502751543
Encompass_Preferred
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter round metal pushbutton switch. It has a black plastic button cap and is rated for 10 amps at 250 volts AC. The pushbutton has silver contacts and includes a legend plate for custom engraving. It mounts into a panel cutout and secures with two threaded studs and hex nuts. This industrial pushbutton switch is commonly used for machine controls and industrial equipment.
Catalog No. 50-275-1543 Supplier Grainger Supplier No. 14A128
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter round metal pushbutton switch. It has a black plastic button cap and is rated for 10 amps at 250 volts AC. The pushbutton has silver contacts and includes a legend plate for custom engraving. It mounts into a panel cutout and secures with two threaded studs and hex nuts. This industrial pushbutton switch is commonly used for machine controls and industrial equipment.