Learn More
Grainger LegendPlate In White/Black Rectangular
Catalog No. 502749316
Encompass_Preferred
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30mm diameter pushbutton switch with a legend plate for panel mounting applications. It has a plastic actuator and body with options for 1, 2, 3, or 4 NO/NC contacts. Contact ratings up to 10A 125/250VAC. Available legends include On/Off, Start/Stop, Forward/Reverse, Local/Remote, and more. Suitable for control panels and industrial machinery. Made in USA.
Catalog No. 50-274-9316 Supplier Grainger Supplier No. 14A117
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30mm diameter pushbutton switch with a legend plate for panel mounting applications. It has a plastic actuator and body with options for 1, 2, 3, or 4 NO/NC contacts. Contact ratings up to 10A 125/250VAC. Available legends include On/Off, Start/Stop, Forward/Reverse, Local/Remote, and more. Suitable for control panels and industrial machinery. Made in USA.