Learn More
Grainger LegendPlate JogFor White/Blk Rectangular
Catalog No. 502753070
Encompass_Preferred
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter pushbutton switch with a legend plate. It has a plastic housing with a black color and a silver legend plate. The switch is rated for 10 amps at 250 volts AC and is suitable for panel mounting applications. It has screw terminal connections and includes a legend plate that can be customized with text or symbols. The pushbutton provides momentary contact.
Catalog No. 50-275-3070 Supplier Grainger Supplier No. 14A125
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter pushbutton switch with a legend plate. It has a plastic housing with a black color and a silver legend plate. The switch is rated for 10 amps at 250 volts AC and is suitable for panel mounting applications. It has screw terminal connections and includes a legend plate that can be customized with text or symbols. The pushbutton provides momentary contact.