Learn More
Grainger LegendPlate JogRun Wht/Blk Rectangular
Catalog No. 502749318
Encompass_Preferred
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter pushbutton operator with a plastic legend plate. It has a black plastic housing and a silver metal button. The pushbutton is rated for 10 amps at 250 volts AC and is commonly used for machine controls and industrial applications. The legend plate allows custom engraving for pushbutton identification. This Eaton pushbutton switch can be panel mounted via the threaded bushing and locknut. It is sold individually.
Catalog No. 50-274-9318 Supplier Grainger Supplier No. 14A121
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter pushbutton operator with a plastic legend plate. It has a black plastic housing and a silver metal button. The pushbutton is rated for 10 amps at 250 volts AC and is commonly used for machine controls and industrial applications. The legend plate allows custom engraving for pushbutton identification. This Eaton pushbutton switch can be panel mounted via the threaded bushing and locknut. It is sold individually.