Learn More
Grainger LegendPlate Low White/Black Rectangular
Catalog No. 502752671
Encompass_Preferred
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter pushbutton switch with a legend plate. It has a plastic housing with a black color and a silver legend plate. The switch is rated for 10 amps at 250 volts AC and is suitable for use as a control switch in industrial applications. It has screw terminal connections and mounts into a 30mm round hole in a panel or enclosure. This Eaton pushbutton switch can be used for machine control or to start and stop electric motors.
Catalog No. 50-275-2671 Supplier Grainger Supplier No. 14A127
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter pushbutton switch with a legend plate. It has a plastic housing with a black color and a silver legend plate. The switch is rated for 10 amps at 250 volts AC and is suitable for use as a control switch in industrial applications. It has screw terminal connections and mounts into a 30mm round hole in a panel or enclosure. This Eaton pushbutton switch can be used for machine control or to start and stop electric motors.