Learn More
Grainger LegendPlate MotorRun Wht/Blk Rectangular
Catalog No. 502748842
Encompass_Preferred
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter round metal pushbutton switch. It is rated for 10 amps at 125/250 volts AC and 14 amps at 125 volts DC. The pushbutton has a black plastic actuator and mounts into a panel cutout. It is commonly used for machine controls and industrial applications. The pushbutton provides momentary contact.
Catalog No. 50-274-8842 Supplier Grainger Supplier No. 14A101
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter round metal pushbutton switch. It is rated for 10 amps at 125/250 volts AC and 14 amps at 125 volts DC. The pushbutton has a black plastic actuator and mounts into a panel cutout. It is commonly used for machine controls and industrial applications. The pushbutton provides momentary contact.