Learn More
Grainger LegendPlate MotorStop Wht/Blk Rectangle
Catalog No. 502749291
Encompass_Preferred
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter pushbutton switch with a plastic actuator and metal mounting plate. It is rated for 10 amps at 250 volts AC and is available in colors including black, red, green, yellow, blue, and white. The switch has silver contacts and includes a legend plate for custom engraving. It can be panel mounted in a 22.5 mm hole and has a depth behind panel of 28.8 mm. The switch is commonly used for machine controls and industrial applications.
Catalog No. 50-274-9291 Supplier Grainger Supplier No. 14A111
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter pushbutton switch with a plastic actuator and metal mounting plate. It is rated for 10 amps at 250 volts AC and is available in colors including black, red, green, yellow, blue, and white. The switch has silver contacts and includes a legend plate for custom engraving. It can be panel mounted in a 22.5 mm hole and has a depth behind panel of 28.8 mm. The switch is commonly used for machine controls and industrial applications.