Learn More
Grainger Eaton Legend Plate, Red, OFF, 30.5 mm, 1 ea
Catalog No. 502748775
Encompass_Preferred
This legend plate is designed for use with the HT800 series of pushbutton switches. Constructed from plastic, this watertight and oiltight accessory provides clear identification for industrial control panels.
  • Legend: OFF
  • Color: Red background with white text
  • Size: 30.5 mm
  • Construction: Watertight and oiltight plastic
  • Compatibility: Designed for the HT800 series
  • Certifications: UL listed and CSA certified
Catalog No. 50-274-8775 Supplier Grainger Supplier No. 14A105
This legend plate is designed for use with the HT800 series of pushbutton switches. Constructed from plastic, this watertight and oiltight accessory provides clear identification for industrial control panels.
  • Legend: OFF
  • Color: Red background with white text
  • Size: 30.5 mm
  • Construction: Watertight and oiltight plastic
  • Compatibility: Designed for the HT800 series
  • Certifications: UL listed and CSA certified