Learn More
Grainger LegendPlate OpenClose Wht/Blk Rectangle
Catalog No. 502749269
Encompass_Preferred
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter pushbutton switch with a legend plate. It has a plastic actuator and body with a silver finish. The switch is rated for 10 amps at 250 volts AC. It has screw terminal connections and mounts into a 30mm round hole. The legend plate allows custom labeling. This pushbutton switch is commonly used for machine controls and industrial applications where a large, heavy duty pushbutton is needed.
Catalog No. 50-274-9269 Supplier Grainger Supplier No. 14A112
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter pushbutton switch with a legend plate. It has a plastic actuator and body with a silver finish. The switch is rated for 10 amps at 250 volts AC. It has screw terminal connections and mounts into a 30mm round hole. The legend plate allows custom labeling. This pushbutton switch is commonly used for machine controls and industrial applications where a large, heavy duty pushbutton is needed.