Learn More
Grainger LegendPlate Reverse Wht/Blk Rectangular
Catalog No. 502748760
Encompass_Preferred
The Grainger 14A106 is a 14 gauge, 3 conductor SJT power cord that is 6 feet long. It has a molded plug on one end and stripped wire leads on the other end. The plug is a NEMA 5-15P for connecting to standard 120 volt outlets. The 3 conductors are black, white, and green for hot, neutral, and ground. The jacket is orange in color. This general purpose power cord can be used for light duty applications up to 15 amps. It's UL listed and CSA certified. The cord is flexible and durable.
Catalog No. 50-274-8760 Supplier Grainger Supplier No. 14A106
The Grainger 14A106 is a 14 gauge, 3 conductor SJT power cord that is 6 feet long. It has a molded plug on one end and stripped wire leads on the other end. The plug is a NEMA 5-15P for connecting to standard 120 volt outlets. The 3 conductors are black, white, and green for hot, neutral, and ground. The jacket is orange in color. This general purpose power cord can be used for light duty applications up to 15 amps. It's UL listed and CSA certified. The cord is flexible and durable.