Learn More
Grainger LegendPlate Safe White/Black Rectangular
Catalog No. 502752263
Encompass_Preferred
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter pushbutton switch with a plastic actuator and metal mounting plate. It is rated for 10 amps at 250 volts AC and is available in colors including black, red, green, yellow, blue, and white. The pushbutton has a momentary contact type with silver alloy contacts. It can be panel mounted using the included nut fastener and has an IP40 ingress protection rating. The overall dimensions are 30 x 41 x 68 mm. This pushbutton switch is commonly used in industrial control panels and machinery.
Catalog No. 50-275-2263 Supplier Grainger Supplier No. 14A130
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter pushbutton switch with a plastic actuator and metal mounting plate. It is rated for 10 amps at 250 volts AC and is available in colors including black, red, green, yellow, blue, and white. The pushbutton has a momentary contact type with silver alloy contacts. It can be panel mounted using the included nut fastener and has an IP40 ingress protection rating. The overall dimensions are 30 x 41 x 68 mm. This pushbutton switch is commonly used in industrial control panels and machinery.