Learn More
Grainger LegendPlate StartStop Wht/Blk Rectangle
Catalog No. 502749315
Encompass_Preferred
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter round metal pushbutton switch. It has a black plastic button cap and is rated for 10 amps at 125/250 volts AC. The pushbutton has silver contacts and includes a legend plate for custom engraving. It mounts into a 1-1/16" diameter panel cutout and has 1/2" NPT conduit entries. This pushbutton can be used for machine controls, industrial equipment, control panels, and other applications requiring a heavy duty 30 mm operator interface. It is UL/CSA certified and made in the USA.
Catalog No. 50-274-9315 Supplier Grainger Supplier No. 14A108
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter round metal pushbutton switch. It has a black plastic button cap and is rated for 10 amps at 125/250 volts AC. The pushbutton has silver contacts and includes a legend plate for custom engraving. It mounts into a 1-1/16" diameter panel cutout and has 1/2" NPT conduit entries. This pushbutton can be used for machine controls, industrial equipment, control panels, and other applications requiring a heavy duty 30 mm operator interface. It is UL/CSA certified and made in the USA.