Learn More
Grainger LegendPlate Test White/Black Rectangular
Catalog No. 502749255
Encompass_Preferred
The Grainger 14A118 is a 14 gauge, 3 conductor SJT power cord that is UL and CSA listed. It features a molded plug on one end and stripped wire leads on the other end, with 3 x 18 AWG wire conductors. The total length of the cord is 15 feet. This general purpose power supply cord is rated for 125V AC and can support up to 15 amps. It can be used for various light industrial and commercial applications requiring an extra long, durable power cord. The flexible SJT jacket stands up well to oils and sunlight. The cord has an operating temperature range of -31°F to 140°F.
Catalog No. 50-274-9255 Supplier Grainger Supplier No. 14A118
The Grainger 14A118 is a 14 gauge, 3 conductor SJT power cord that is UL and CSA listed. It features a molded plug on one end and stripped wire leads on the other end, with 3 x 18 AWG wire conductors. The total length of the cord is 15 feet. This general purpose power supply cord is rated for 125V AC and can support up to 15 amps. It can be used for various light industrial and commercial applications requiring an extra long, durable power cord. The flexible SJT jacket stands up well to oils and sunlight. The cord has an operating temperature range of -31°F to 140°F.