Learn More
Grainger LegendPlate Trip White/Black Rectangular
Catalog No. 502749270
Encompass_Preferred
The Grainger 14A114 is a 14 gauge, 3 conductor SJT power cord that is UL and CSA listed. It features a molded plug on one end and stripped wire leads on the other end, with 3 x 14 AWG conductors. The cord is 15 feet long, black in color, and rated for 300 volts. The molded plug has a NEMA 5-15P configuration for connecting to standard 120 volt outlets. This general purpose power cord is well suited for many applications requiring a flexible 15 foot cord to connect equipment to an outlet.
Catalog No. 50-274-9270 Supplier Grainger Supplier No. 14A114
The Grainger 14A114 is a 14 gauge, 3 conductor SJT power cord that is UL and CSA listed. It features a molded plug on one end and stripped wire leads on the other end, with 3 x 14 AWG conductors. The cord is 15 feet long, black in color, and rated for 300 volts. The molded plug has a NEMA 5-15P configuration for connecting to standard 120 volt outlets. This general purpose power cord is well suited for many applications requiring a flexible 15 foot cord to connect equipment to an outlet.