Learn More
Grainger LegendPlate White/Red/Black Rectangular
Catalog No. 502753608
Encompass_Preferred
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter round metal pushbutton switch. It has a black plastic button cap and mounts into a panel cutout. The pushbutton has 2 NO contacts and is rated for 10 amps at 250 volts AC. It is commonly used for machine controls and industrial applications where a rugged metal pushbutton is needed. The pushbutton has a threaded bushing for secure mounting into a panel.
Catalog No. 50-275-3608 Supplier Grainger Supplier No. 14A134
The Eaton Legend Plate 30 mm Panel Mounting Pushbutton is a 30 mm diameter round metal pushbutton switch. It has a black plastic button cap and mounts into a panel cutout. The pushbutton has 2 NO contacts and is rated for 10 amps at 250 volts AC. It is commonly used for machine controls and industrial applications where a rugged metal pushbutton is needed. The pushbutton has a threaded bushing for secure mounting into a panel.