Learn More
Biolegend LEGENDplex™ Human Inflammation Panel 1 (13-plex) w/VbP
Catalog No. NC1901280
Encompass_Preferred
LEGENDplex™ Human Inflammation Panel 1 (13-plex) w/VbP Reactivity: Human; Apps: Multiplex - Panel; Size: 100 tests
Catalog No. NC1901280 Supplier Biolegend Supplier No. 740809
LEGENDplex™ Human Inflammation Panel 1 (13-plex) w/VbP Reactivity: Human; Apps: Multiplex - Panel; Size: 100 tests