Learn More
Biolegend LEGENDplex™ HU Fibrinolysis Panel (5-plex) w/ VbP
Catalog No. 502088349
Encompass_Preferred
LEGENDplex™ HU Fibrinolysis Panel (5-plex) w/ VbP Reactivity: Human; Apps: Multiplex - Panel; Size: 100 tests
Catalog No. 50-208-8349 Supplier Biolegend Supplier No. 740761
LEGENDplex™ HU Fibrinolysis Panel (5-plex) w/ VbP Reactivity: Human; Apps: Multiplex - Panel; Size: 100 tests