Learn More
Biolegend LEGENDplex NHP Th Cytokine Panel V02 (12-plex) w/ FP Reactivity Non-Human Primate Apps Multiplex - Panel Size 100 tests
Catalog No. 5000496173
Encompass_Preferred
LEGENDplex NHP Th Cytokine Panel V02 (12-plex) w/ FP Reactivity Non-Human Primate Apps Multiplex - Panel Size 100 tests
Catalog No. 50-004-96173 Supplier Biolegend Supplier No. 741633
LEGENDplex NHP Th Cytokine Panel V02 (12-plex) w/ FP Reactivity Non-Human Primate Apps Multiplex - Panel Size 100 tests