Learn More
National Safety Apparel™ Legging 5 oz. Carbon Kevlar Lined With 6 oz. Omni Quilted
SDP
Catalog No. 170012044 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-2044 Large
17-001-2045 X-Large
17-001-2046 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-001-2044 Supplier National Safety Apparel™ Supplier No. L05NPQHLTLG
Add to Cart
Add to Cart
Specifications
Size Large
Gender Unisex
Color Olive
Ankle Style Open
Seam Type Stitched
Disposable Reusable
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Sterility Non-sterile