Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Legionella Latex Test

Catalog No. DR0802M Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Legionella Latex Test
Serogroup 1 Test Reagent
Serogroup 2-14 Test Reagent
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
DR0802M Serogroup 2-14 Test Reagent
OXDR0800M Legionella Latex Test
OXDR0801M Serogroup 1 Test Reagent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DR0802M Supplier Thermo Scientific™ Supplier No. DR0802M
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapidly confirm predominant Legionella serogroups grown on selective media by latex agglutination.

Rapidly differentiate and confirm the identity of predominant Legionella serogroups grown on selective media by latex agglutination using Thermo Scientific™ Legionella Latex Test.

Use this latex agglutination test for the identification of predominant Legionella species grown on plated media from patients with suspected Legionellosis or from environmental sources. The test allows separate identification of Legionella pneumophila serogroup 1 and serogroups 2-14 and detection of seven other Legionella species which have been implicated in human disease.

Blue latex particles sensitised with antibody agglutinate in the presence of specific Legionella cell wall antigens to form visible clumps.

The following products are available for use with the Legionella Latex Test and are sold separately:

Legionella Pneumophila Serogroup 1 Test Reagent (Part No. DR0801M)

Legionella Pneumophila Serogroup 2-14 Test Reagent (Part No. DR0802M)

Legionella Species Test Reagent (Part No. DR0800M)—Legionella longbeachae 1 & 2, Legionella bozemanii 1 & 2, Legionella dumoffii, Legionella gormanii, Legionella jordanis, Legionella micdadei and Legionella anisa

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Serogroup 2-14 Test Reagent
Detectable Analytes Legionella
Sample Type Environmental Source
Type Legionella Latex Test Kit
Quantity 50 Tests/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.