Learn More
ATCC Legionella longbeachae; Long Beach 4
Catalog No. 502384457
Encompass
Legionella longbeachae strain Long Beach 4 [NCTC 11477] is a whole-genome sequenced bacterial type strain that was isolated from a human lung. This bacterial culture can be used in bioinformatics.
Catalog No. 50-238-4457 Supplier ATCC Supplier No. 33462
Add to Cart
Add to Cart
Legionella longbeachae strain Long Beach 4 [NCTC 11477] is a whole-genome sequenced bacterial type strain that was isolated from a human lung. This bacterial culture can be used in bioinformatics.