Learn More
ATCC Legionella pneumophila; Philadelphia-1; genomic DNA
Catalog No. 502384099
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Genomic DNA isolated from Legionella pneumophila subsp. pneumophila strain Philadelphia-1 (ATCC 33152). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-4099 Supplier ATCC Supplier No. 33152D5
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Genomic DNA isolated from Legionella pneumophila subsp. pneumophila strain Philadelphia-1 (ATCC 33152). This whole-genome sequenced product can be used in PCR and other molecular biology applications.