Learn More
ATCC Legionella pneumophila; subsp. pneumophila
Supplier:  ATCC 33152
Encompass
Legionella pneumophila subsp. pneumophila strain Philadelphia-1 was isolated from a human lung. This whole-genome sequenced bacterial type strain can be used in media testing, emerging infectious disease research, and respiratory disease research.
Catalog No. 50-238-2651