Learn More
ATCC Corynebacterium jeikeium
Supplier:  ATCC 43734
Encompass
Corynebacterium jeikeium strain NCTC 11913 is a bacterial type strain isolated from blood culture. This strain is propagated aerobically at 37°C.
Catalog No. 50-238-1333