Learn More
ATCC Cedecea neteri; Strain CDC 0621-75
Catalog No. 502381616
Encompass
Cedecea neteri strain CDC 0621-75 was isolated from a human foot in California. This bacterial type strain is propagated aerobically on trypticase soy medium.
Catalog No. 50-238-1616 Supplier ATCC Supplier No. 33855
Cedecea neteri strain CDC 0621-75 was isolated from a human foot in California. This bacterial type strain is propagated aerobically on trypticase soy medium.