Learn More
Seracare Life Sciences Inc LEGIONLLA PNEUMOPHILA POS CTRL
Catalog No. 5067460
Encompass
Heat-killed Legionaella pneumophila cells in dextran solution. Lot specific cell counts can be obtained by requesting a Certificate of Analysis or by calling Technical Services with the specific lot number
This item is not returnable. View return policy
Catalog No. 50-674-60 Supplier Seracare Life Sciences Inc Supplier No. 53700005
Heat-killed Legionaella pneumophila cells in dextran solution. Lot specific cell counts can be obtained by requesting a Certificate of Analysis or by calling Technical Services with the specific lot number