Learn More
Grainger LegndPlate ForOffRev Wht/Blk Rectangular
Catalog No. 502749290
Encompass_Preferred
The Grainger 14A104 is a 14 gauge, type A, class 1, 4 conductor SJOOW portable power cable. It is 600 volt rated with 90°C insulation. The cable has an oil resistant outer PVC jacket and 4 color coded THHN stranded copper conductors. This general purpose portable power cable is flexible for use in applications like construction sites, stage lighting, machine wiring, and diesel generator hookups. The 14 gauge allows it to carry up to 15 amps. This 100 foot reel of SJOOW cable from Grainger provides durable and flexible power connections.
Catalog No. 50-274-9290 Supplier Grainger Supplier No. 14A104
The Grainger 14A104 is a 14 gauge, type A, class 1, 4 conductor SJOOW portable power cable. It is 600 volt rated with 90°C insulation. The cable has an oil resistant outer PVC jacket and 4 color coded THHN stranded copper conductors. This general purpose portable power cable is flexible for use in applications like construction sites, stage lighting, machine wiring, and diesel generator hookups. The 14 gauge allows it to carry up to 15 amps. This 100 foot reel of SJOOW cable from Grainger provides durable and flexible power connections.