Learn More
Medchemexpress LLC Legumain inhibitor 1 | 2361157-34-6 | 98.3% | 100 MG
SDP
Catalog No. 5000339321
Encompass_Preferred
Legumain inhibitor 1 is a potent and selective Legumain inhibitor with an IC50 of 3.6 nM. It is suitable for cancer research.
  • Potent and selective Legumain inhibitor
  • IC50 of 3.6 nM
  • Suitable for cancer research
  • Demonstrates high microsomal stability in vitro
  • Half-lives exceeding 8 hours in mouse and human MICs
Catalog No. 50-003-39321 Supplier Medchemexpress LLC Supplier No. HY128617100MG
Legumain inhibitor 1 is a potent and selective Legumain inhibitor with an IC50 of 3.6 nM. It is suitable for cancer research.
  • Potent and selective Legumain inhibitor
  • IC50 of 3.6 nM
  • Suitable for cancer research
  • Demonstrates high microsomal stability in vitro
  • Half-lives exceeding 8 hours in mouse and human MICs