Learn More
Medchemexpress LLC Lei-101 | 2250025-91-1 | 99.4% | 508.99 g·mol⁻¹ | C23H26ClFN4O4S | 10MG
SDP
Catalog No. 5000238942
Encompass_Preferred
LEI-101 is a potent, selective, orally bioavailable cannabinoid CB2 receptor agonist used in research to investigate CB2-mediated signaling and pharmacology. It exhibits approximately 100-fold selectivity for CB2 over CB1, with a pEC50 of about 8 for human CB2, low hERG affinity, and is supplied as a high-purity white to off-white solid suitable for in vitro and in vivo studies.
  • High purity (99.4%).
  • Potent CB2 agonist (pEC50 ≈ 8 for hCB2).
  • Approximately 100-fold selectivity for CB2 over CB1.
  • Low hERG binding (pKi < 4).
  • Soluble in DMSO (≈116.67 mg/mL).
  • Stable when stored sealed and kept away from moisture; in solvent: -80°C (6 months), -20°C (1 month).
  • Provided in milligram-scale quantities for research use.
Catalog No. 50-002-38942 Supplier Medchemexpress LLC Supplier No. HY124283A10MG
LEI-101 is a potent, selective, orally bioavailable cannabinoid CB2 receptor agonist used in research to investigate CB2-mediated signaling and pharmacology. It exhibits approximately 100-fold selectivity for CB2 over CB1, with a pEC50 of about 8 for human CB2, low hERG affinity, and is supplied as a high-purity white to off-white solid suitable for in vitro and in vivo studies.
  • High purity (99.4%).
  • Potent CB2 agonist (pEC50 ≈ 8 for hCB2).
  • Approximately 100-fold selectivity for CB2 over CB1.
  • Low hERG binding (pKi < 4).
  • Soluble in DMSO (≈116.67 mg/mL).
  • Stable when stored sealed and kept away from moisture; in solvent: -80°C (6 months), -20°C (1 month).
  • Provided in milligram-scale quantities for research use.