Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lei-101 hydrochloride | 2250025-91-1 | 99.4% | 508.99 | C23H26ClFN4O4S | 100MG
SDP

Catalog No. 5000238523
Encompass_Preferred

LEI-101 is a potent, selective cannabinoid CB2 receptor agonist used for biochemical and pharmacological research. It shows high selectivity for CB2 over CB1, is orally bioavailable, and is supplied as a solid for laboratory studies. Intended for research use only (not for human use).

  • Selective CB2 receptor agonist with approximately 100-fold selectivity over CB1.
  • Orally bioavailable, suitable for in vivo pharmacology studies.
  • High purity (99.35%).
  • Molecular weight 508.99 and formula C23H26ClFN4O4S.
  • Soluble in DMSO (approx. 116.7 mg/mL).
  • Intended for laboratory research use only, not for human consumption.

Catalog No. 50-002-38523 Supplier Medchemexpress LLC Supplier No. HY124283A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LEI-101 is a potent, selective cannabinoid CB2 receptor agonist used for biochemical and pharmacological research. It shows high selectivity for CB2 over CB1, is orally bioavailable, and is supplied as a solid for laboratory studies. Intended for research use only (not for human use).

  • Selective CB2 receptor agonist with approximately 100-fold selectivity over CB1.
  • Orally bioavailable, suitable for in vivo pharmacology studies.
  • High purity (99.35%).
  • Molecular weight 508.99 and formula C23H26ClFN4O4S.
  • Soluble in DMSO (approx. 116.7 mg/mL).
  • Intended for laboratory research use only, not for human consumption.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.