Learn More
Medchemexpress LLC Lei 101 hydrochloride | 2250025-91-1 | 508.99 g/mol | 50mg
SDP
Catalog No. 5000238230
Encompass_Preferred
LEI-101 is a potent selective and orally bioavailable cannabinoid CB2 receptor agonist with a pEC50 of 8 for hCB2 and a pKi of less than 4 for hERG LEI-101 is 100-fold more potent in binding to CB2 receptors than to CB1 receptors[1][2]
Catalog No. 50-002-38230 Supplier Medchemexpress LLC Supplier No. HY124283A50MG
LEI-101 is a potent selective and orally bioavailable cannabinoid CB2 receptor agonist with a pEC50 of 8 for hCB2 and a pKi of less than 4 for hERG LEI-101 is 100-fold more potent in binding to CB2 receptors than to CB1 receptors[1][2]