Learn More
Medchemexpress LLC HY-131181 5mg Medchemexpress, LEI-401 CAS:2393840-15-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1311815MG
Encompass_Preferred
Medchemexpress, HY-131181 5mg LEI-401 CAS:2393840-15-6 LEI-401 is a first-in-class, selective, and CNS-active NAPE-PLD (N-acylphosphatidylethanolamine phospholipase D) inhibitor, with an IC50 of 27 nM. LEI-401 modulates emotional behavior in mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0252