Learn More
Medchemexpress LLC Lei110 | 2313525-90-3 | 99.7% | 456.46 | C25H23F3N2O3 | 25MG
SDP
Catalog No. 5000235078
Encompass_Preferred
LEI110 is a small-molecule inhibitor of the HRASLS family of thiol hydrolases, including PLA2G16. It is cell-permeable and intended for research use in biochemical and cellular assays to probe phospholipase-related pathways; handle according to the safety data sheet and laboratory best practices.
  • Potent inhibitor of HRASLS/PLA2G16 suitable for biochemical assays.
  • Cell-permeable for use in cellular studies.
  • High purity confirmed by HPLC (99.72%).
  • Characterized by COA and SDS with formula C25H23F3N2O3 and molecular weight 456.46.
  • Soluble in DMSO and recommended for storage under desiccated cold conditions.
  • Available in small mass pack sizes appropriate for research applications.
Catalog No. 50-002-35078 Supplier Medchemexpress LLC Supplier No. HY12525425MG
LEI110 is a small-molecule inhibitor of the HRASLS family of thiol hydrolases, including PLA2G16. It is cell-permeable and intended for research use in biochemical and cellular assays to probe phospholipase-related pathways; handle according to the safety data sheet and laboratory best practices.
  • Potent inhibitor of HRASLS/PLA2G16 suitable for biochemical assays.
  • Cell-permeable for use in cellular studies.
  • High purity confirmed by HPLC (99.72%).
  • Characterized by COA and SDS with formula C25H23F3N2O3 and molecular weight 456.46.
  • Soluble in DMSO and recommended for storage under desiccated cold conditions.
  • Available in small mass pack sizes appropriate for research applications.