Learn More
ATCC Leibovitz's L-15 Medium, 500mL
Supplier:  ATCC 302008/DEL
Encompass
It is modified to contain 2 mM L-glutamine and no sodium bicarbonate. This formulation is designed to be used without additional CO2 in the atmosphere of cell cultures. For use by FOR-PROFIT companies only; NON-PROFIT companies use 50188273NP. New ATCC customers are required to register with ATCC prior to purchase. The application can be found at https://www.atcc.org/en/My_Profile/Register.aspx
Catalog No. 50-188-273FP