Learn More
ATCC Leibovitz's L-15 Medium
Catalog No. 502383972
Encompass
Leibovitz's L-15 Medium contains 2 mM L-glutamine and no sodium bicarbonate. This formulation is designed to be used without additional CO2 in the atmosphere of cell cultures. It has applications for cell culture, cell growth, and viability.
Catalog No. 50-238-3972 Supplier ATCC Supplier No. 302008
Leibovitz's L-15 Medium contains 2 mM L-glutamine and no sodium bicarbonate. This formulation is designed to be used without additional CO2 in the atmosphere of cell cultures. It has applications for cell culture, cell growth, and viability.