Learn More
Leica Biosystems LEICA 3P O-FIX 1 GALLON
Catalog No. NC1382585
Encompass_Preferred
O-fix is a tissue fixative formulated with alcohol, formalin and acetic acid. Although it can be used to fix all types and sizes of tissue specimens, it is especially helpful in highlighting lymph nodes by turning them an opaque white.
Catalog No. NC1382585 Supplier Leica Biosystems Supplier No. 3800676
O-fix is a tissue fixative formulated with alcohol, formalin and acetic acid. Although it can be used to fix all types and sizes of tissue specimens, it is especially helpful in highlighting lymph nodes by turning them an opaque white.