Learn More
Leica Biosystems Leica Biosystems 7 mL PD-L1 (CAL10) RUO BOND RTU Primary Antibody
Catalog No. NC3863819
Encompass
Leica Biosystems 7 mL PD-L1 (CAL10) RUO BOND RTU Primary Antibody is a ready-to-use rabbit monoclonal reagent formulated for immunohistochemistry on automated platforms. Supplied in a buffered carrier solution with preservative, it is optimized for the detection of human PD-L1 in research applications and is not for diagnostic use.
  • Ready-to-use 7 mL format for automated staining workflows.
  • Rabbit monoclonal antibody, clone CAL10, for PD-L1 detection.
  • Formulated in Tris-buffered saline with carrier protein for stability.
  • Contains preservative to support reagent integrity during storage.
  • Intended for research use only; not for diagnostic procedures.
  • Documentation and instructions for use available from the manufacturer.
Catalog No. NC3863819 Supplier Leica Biosystems Supplier No. PA0456RUO
Leica Biosystems 7 mL PD-L1 (CAL10) RUO BOND RTU Primary Antibody is a ready-to-use rabbit monoclonal reagent formulated for immunohistochemistry on automated platforms. Supplied in a buffered carrier solution with preservative, it is optimized for the detection of human PD-L1 in research applications and is not for diagnostic use.
  • Ready-to-use 7 mL format for automated staining workflows.
  • Rabbit monoclonal antibody, clone CAL10, for PD-L1 detection.
  • Formulated in Tris-buffered saline with carrier protein for stability.
  • Contains preservative to support reagent integrity during storage.
  • Intended for research use only; not for diagnostic procedures.
  • Documentation and instructions for use available from the manufacturer.