Learn More
Leica Biosystems Leica Biosystems Elution Solution for multiplex IHC, ready-to-use 7 mL (AR9471)
Catalog No. NC3903976
Encompass
Elution Solution is a ready-to-use reagent formulated to remove residual antibody from preceding detection layers during sequential multiplex immunohistochemistry, preventing crosstalk and ensuring staining specificity on automated research stainers.
  • Removes residual antibody from prior detection layers to prevent crosstalk.
  • Ready-to-use, automated configuration for simplified workflow.
  • Suitable for multiplex chromogenic staining protocols.
  • Research use only (RUO) formulation for laboratory applications.
  • Pack volume provides approximately seven tests (7 mL).
Catalog No. NC3903976 Supplier Leica Biosystems Supplier No. AR9471
Elution Solution is a ready-to-use reagent formulated to remove residual antibody from preceding detection layers during sequential multiplex immunohistochemistry, preventing crosstalk and ensuring staining specificity on automated research stainers.
  • Removes residual antibody from prior detection layers to prevent crosstalk.
  • Ready-to-use, automated configuration for simplified workflow.
  • Suitable for multiplex chromogenic staining protocols.
  • Research use only (RUO) formulation for laboratory applications.
  • Pack volume provides approximately seven tests (7 mL).