Learn More
Leica Biosystems Leica Biosystems HRP Yellow Chromogen (DC9462), Ready-to-Use, 50 tests
Catalog No. NC3916370
Encompass
Leica Biosystems HRP Yellow Chromogen (DC9462) is a ready-to-use, research-use-only chromogenic substrate that produces a yellow reaction product in the presence of horseradish peroxidase (HRP) for use in immunohistochemistry (IHC) and in situ hybridization (ISH). It is formulated for reliable single-plex or multiplex staining workflows and is supplied in a pack size suitable for approximately 50 tests.
  • Produces a yellow reaction product with HRP
  • Ready-to-use formulation for streamlined workflow
  • Suitable for single or multiplex IHC and ISH staining
  • Compatible with automated research stainers
  • Research use only; not for diagnostic procedures
  • Typical pack size: approximately 50 tests
Catalog No. NC3916370 Supplier Leica Biosystems Supplier No. DC9462
Leica Biosystems HRP Yellow Chromogen (DC9462) is a ready-to-use, research-use-only chromogenic substrate that produces a yellow reaction product in the presence of horseradish peroxidase (HRP) for use in immunohistochemistry (IHC) and in situ hybridization (ISH). It is formulated for reliable single-plex or multiplex staining workflows and is supplied in a pack size suitable for approximately 50 tests.
  • Produces a yellow reaction product with HRP
  • Ready-to-use formulation for streamlined workflow
  • Suitable for single or multiplex IHC and ISH staining
  • Compatible with automated research stainers
  • Research use only; not for diagnostic procedures
  • Typical pack size: approximately 50 tests