Learn More
Leica Biosystems Leica Biosystems Movable Shelf for Cryostat (fits CM1520)
Catalog No. NC3815157
Encompass
The Leica Biosystems movable shelf is an adjustable internal shelf designed for use inside compatible Leica cryostats, providing added workspace and storage within the instrument chamber.
  • Provides adjustable storage and workspace inside the cryostat chamber.
  • Movable design allows flexible positioning to suit different workflows.
  • Removable for easy cleaning and maintenance.
  • Helps organize samples and accessories within the instrument.
Catalog No. NC3815157 Supplier Leica Biosystems Supplier No. 14049146750
The Leica Biosystems movable shelf is an adjustable internal shelf designed for use inside compatible Leica cryostats, providing added workspace and storage within the instrument chamber.
  • Provides adjustable storage and workspace inside the cryostat chamber.
  • Movable design allows flexible positioning to suit different workflows.
  • Removable for easy cleaning and maintenance.
  • Helps organize samples and accessories within the instrument.