Learn More
Leica Biosystems Leica Biosystems Replacement O-ring for Instruments, 28 x 3 mm (FKM)
Supplier:  Leica Biosystems 14025339635
Encompass
Leica Biosystems part 14025339635 is an O-ring sized 28 x 3 mm, made from FKM (fluorocarbon rubber). It is supplied as a service spare for instrument assemblies and maintenance kits, providing a resilient seal for applications where chemical and temperature resistance are important.
  • Made from FKM (fluorocarbon rubber) for chemical and temperature resistance.
  • Dimensions: 28 x 3 mm (ID x cross-section).
  • Designed as a replacement sealing O-ring for instrument assemblies.
  • Suitable for sealing fluid and gas interfaces.
Catalog No. NC3844368