Learn More
Leica Biosystems Leica BOND Printer Ribbon & Labels Cxi, 6-pack (for BOND slide labeler/printer)
Catalog No. NC3866659
Encompass
Leica BOND Printer Ribbon & Labels Cxi (6-pack) provides replacement printer ribbon and pre-cut slide labels for Leica BOND slide labeler/printer systems. The kit supplies multiple ribbon and label rolls to support slide identification and specimen tracking workflows in histology and immunohistochemistry laboratories.
  • Contains six replacement ribbon and label kits.
  • Compatible with BOND slide labeler/printer systems.
  • Labels adhere to slides for secure identification.
  • Limited resistance to common solvents; validate label use in your laboratory.
  • Label reagent stability: xylene, toluene, acetone - < 4 hours; water and aqueous reagents - < 6 hours.
  • Sold as a consumable for slide labeling and barcode printing workflows.
Catalog No. NC3866659 Supplier Leica Biosystems Supplier No. S21.4610
Add to Cart
Add to Cart
Request a Quote
Leica BOND Printer Ribbon & Labels Cxi (6-pack) provides replacement printer ribbon and pre-cut slide labels for Leica BOND slide labeler/printer systems. The kit supplies multiple ribbon and label rolls to support slide identification and specimen tracking workflows in histology and immunohistochemistry laboratories.
  • Contains six replacement ribbon and label kits.
  • Compatible with BOND slide labeler/printer systems.
  • Labels adhere to slides for secure identification.
  • Limited resistance to common solvents; validate label use in your laboratory.
  • Label reagent stability: xylene, toluene, acetone - < 4 hours; water and aqueous reagents - < 6 hours.
  • Sold as a consumable for slide labeling and barcode printing workflows.