Learn More
Leica Biosystems LEICA BOND REAGENT TRAY
Catalog No. NC1336148
Encompass
Additional BOND Reagent Trays let laboratories setup reagents for upcoming runs while other reagent trays are in use.This reduces setup delays and improves laboratory workflow.
Catalog No. NC1336148 Supplier Leica Biosystems Supplier No. S21.1003
Additional BOND Reagent Trays let laboratories setup reagents for upcoming runs while other reagent trays are in use.This reduces setup delays and improves laboratory workflow.