Learn More
Leica Biosystems Leica ChromoPlex III Triple Detection Kit for BOND RX/RXm (100 tests)
Catalog No. NC3831983
Encompass
ChromoPlex III Triple Detection RUO is a three-plex chromogenic IHC detection kit designed for automated staining on BOND RX and RXm platforms. The kit simplifies multiplex workflows by combining pre-optimized reagents and protocols to visualize up to three markers on a single tissue section.
  • Enables three-plex chromogenic detection on a single tissue section.
  • Pre-optimized reagents and protocols to reduce assay variability.
  • Biotin-free polymeric HRP and polymeric AP detection chemistry.
  • Includes stable brown, red, and green chromogens for distinct staining.
  • Configured as 16 reagent containers organized into two trays.
  • Supplies sufficient reagents for 100 tests per kit.
  • Designed for automated use to minimize hands-on time.
  • Intended for research use only.
Catalog No. NC3831983 Supplier Leica Biosystems Supplier No. 8123767
Add to Cart
Add to Cart
Request a Quote
ChromoPlex III Triple Detection RUO is a three-plex chromogenic IHC detection kit designed for automated staining on BOND RX and RXm platforms. The kit simplifies multiplex workflows by combining pre-optimized reagents and protocols to visualize up to three markers on a single tissue section.
  • Enables three-plex chromogenic detection on a single tissue section.
  • Pre-optimized reagents and protocols to reduce assay variability.
  • Biotin-free polymeric HRP and polymeric AP detection chemistry.
  • Includes stable brown, red, and green chromogens for distinct staining.
  • Configured as 16 reagent containers organized into two trays.
  • Supplies sufficient reagents for 100 tests per kit.
  • Designed for automated use to minimize hands-on time.
  • Intended for research use only.