Learn More
Leica Biosystems Leica CM1860 UV Cryostat
Catalog No. NC3815156
Encompass
The Leica CM1860 UV Cryostat is a clinical histopathology cryostat engineered for fast, reliable frozen-section preparation in high-throughput laboratories. It delivers consistent section quality, ergonomic operation, and optional contamination-control features for safer workflow.
  • Fast frozen-section throughput with consistent section quality.
  • Quick-freeze shelf reaches down to -40 °C for rapid specimen freezing.
  • Cryochamber temperature range 0 °C to -35 °C for routine processing.
  • Eight freezing stations and two Peltier elements for flexible sample handling.
  • Automatic hot gas defrost for the cryochamber; manual hot gas defrost for the quick-freeze shelf.
  • AgProtect nanosilver coating reduces touch-surface contamination.
  • Optional certified UVC disinfection with selectable 30 or 180 minute cycles.
Catalog No. NC3815156 Supplier Leica Biosystems Supplier No. 1491860UVUS
Add to Cart
Add to Cart
Request a Quote
The Leica CM1860 UV Cryostat is a clinical histopathology cryostat engineered for fast, reliable frozen-section preparation in high-throughput laboratories. It delivers consistent section quality, ergonomic operation, and optional contamination-control features for safer workflow.
  • Fast frozen-section throughput with consistent section quality.
  • Quick-freeze shelf reaches down to -40 °C for rapid specimen freezing.
  • Cryochamber temperature range 0 °C to -35 °C for routine processing.
  • Eight freezing stations and two Peltier elements for flexible sample handling.
  • Automatic hot gas defrost for the cryochamber; manual hot gas defrost for the quick-freeze shelf.
  • AgProtect nanosilver coating reduces touch-surface contamination.
  • Optional certified UVC disinfection with selectable 30 or 180 minute cycles.