Learn More
Leica Biosystems Leica CV Ultra Mounting Media, 1 x 250 mL
Catalog No. NC3808596
Encompass
Leica CV Ultra Mounting Media is a non-solvent coverslipping reagent formulated to replace xylene and toluene in histology and cytology workflows. It is compatible with manual and robotic coverslippers and designed for flexible mounting without hazardous organic solvents.
  • Replaces xylene and toluene and conventional mounting media.
  • Compatible with common robotic coverslippers and manual coverslipping procedures.
  • Does not contain dangerous organic solvents.
  • Flexible use with xylene- and toluene-based workflows.
  • Supplied in a 1 x 250 mL bottle.
Catalog No. NC3808596 Supplier Leica Biosystems Supplier No. 14070937891
Leica CV Ultra Mounting Media is a non-solvent coverslipping reagent formulated to replace xylene and toluene in histology and cytology workflows. It is compatible with manual and robotic coverslippers and designed for flexible mounting without hazardous organic solvents.
  • Replaces xylene and toluene and conventional mounting media.
  • Compatible with common robotic coverslippers and manual coverslipping procedures.
  • Does not contain dangerous organic solvents.
  • Flexible use with xylene- and toluene-based workflows.
  • Supplied in a 1 x 250 mL bottle.