Learn More
Hach Company Leica DM500 Comp. Microscope
Catalog No. 5000207616 Shop All Hach Company Products
Encompass
Leica DM500 Compound MicroscopeThe Leica DM500 microscopes provide superior image quality along with durability and a practical design that truly meets your needs The brightfield achromatic model offers 4x 10x 40x and 100x plan objectives This model comes with led illumination that offers a minimum of 25000 hours of daylight color temperature illumination (the bulbs do not burn out at 25000 hours but dim in brightness) and many other Leica unique features such as Agtreat which helps prevent the spread of surface bacteria The universal power source allows easy conversion when used with the local country power cord Power supply 100-240V 50/60 Hz with standard U S power cord Standard Euro power cord available and sold separately
Catalog No. 50-002-07616 Supplier Hach Company Supplier No. 2947050
Leica DM500 Compound MicroscopeThe Leica DM500 microscopes provide superior image quality along with durability and a practical design that truly meets your needs The brightfield achromatic model offers 4x 10x 40x and 100x plan objectives This model comes with led illumination that offers a minimum of 25000 hours of daylight color temperature illumination (the bulbs do not burn out at 25000 hours but dim in brightness) and many other Leica unique features such as Agtreat which helps prevent the spread of surface bacteria The universal power source allows easy conversion when used with the local country power cord Power supply 100-240V 50/60 Hz with standard U S power cord Standard Euro power cord available and sold separately