Learn More
Electron Microscopy Sciences Applied Physics LaB6 Cathode Leica/LEO Cambridge SMVM
SDP
Catalog No. 50949847
Encompass
Lanthanum hexaboride (LaB6) and cerium hexaboride (CeB6) cathodes are ideal for many small spot size applications such as SEM, TEM, surface analysis and metrology, and for high current applications such as microwave tubes, lithography, electron-beam welders, X-ray sources and free electron lasers. Greater brightness from low work function yielding higher currents at lower temps than tungsten. Cambridge SMVM base.
Type: LaB6
Angle: 90°
Tips: 16 mm
Catalog No. 50-949-847 Supplier Electron Microscopy Sciences Supplier No. 80929LP
Add to Cart
Add to Cart
Lanthanum hexaboride (LaB6) and cerium hexaboride (CeB6) cathodes are ideal for many small spot size applications such as SEM, TEM, surface analysis and metrology, and for high current applications such as microwave tubes, lithography, electron-beam welders, X-ray sources and free electron lasers. Greater brightness from low work function yielding higher currents at lower temps than tungsten. Cambridge SMVM base.
Type: LaB6
Angle: 90°
Tips: 16 mm